Learn More
Strem, An Ascensus Company Me4 t-BuXPhos min. 98% 250mg
SDP
Supplier:  Strem, An Ascensus Company 151051250MG
Encompass
CAS# 857356-94-6. 250mg. 2-Di-t-butylphosphino-3,4,5,6-tetramethyl-2',4',6'-tri-i-propyl)-1,1'-biphenyl, min. 98% Me4 t-BuXPhos. MFCD09038436. Molecular Weight: 480.75. Molecular Formula: C33H53P. Color/form: white microxtl. Strem# 15-1051. http://www.strem.com/catalog/v/15-1051/
Catalog No. 50-901-10048