Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-(Dibenzylamino)-6'-(diethylamino)fluoran 98.0+%, TCI America™
SDP

Catalog No. D320425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D320425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D320425G Supplier TCI America Supplier No. D320425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 34372-72-0
Molecular Formula C38H34N2O3
Molecular Weight (g/mol) 566.701
InChI Key DXYZIGZCEVJFIX-UHFFFAOYSA-N
PubChem CID 118663
IUPAC Name 2'-(dibenzylamino)-6'-(diethylamino)spiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES CCN(CC)C1=CC2=C(C=C1)C3(C4=CC=CC=C4C(=O)O3)C5=C(O2)C=CC(=C5)N(CC6=CC=CC=C6)CC7=CC=CC=C7

Specifications

Melting Point 174°C
Quantity 25 g
Formula Weight 566.70
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′-(Dibenzylamino)-6′-(diethylamino)fluoran
EINECSNumber (5)-3626
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.