Learn More
Strem, An Ascensus Company CAS# 1398565-95-1. 250mg. [2-Dicyclohexylphosphino-3-methoxy-N-methyl-N-phenylbenzenamine, 98% Zheda-Phos. MFCD28400096
SDP
Supplier:  Strem, An Ascensus Company 156528250MG
Encompass
CAS# 1398565-95-1. 250mg. [2-Dicyclohexylphosphino-3-methoxy-N-methyl-N-phenylbenzenamine, 98% Zheda-Phos. MFCD28400096. Molecular Weight: 409.54. Molecular Formula: C26H36NOP. Color/form: white pwdr. Strem# 15-6528. http://www.strem.com/catalog/v/15-6528
Catalog No. 50-138-5428