Learn More
Strem, An Ascensus Company CAS# 1848244-75-6. 250mg. 2-Dicyclohexylphosphino-2'-methoxy-4',6'-di-t-butyl-1,1'-biphenyl, min. 98% VPhos.
SDP
Supplier:  Strem, An Ascensus Company 151105250MG
Encompass
CAS# 1848244-75-6. 250mg. 2-Dicyclohexylphosphino-2'-methoxy-4',6'-di-t-butyl-1,1'-biphenyl, min. 98% VPhos. . Molecular Weight: 492.72. Molecular Formula: C33H49OP. Color/form: white to off-white solid Strem# 15-1105. http://www.strem.com/catalog/v/15-1105/
Catalog No. 50-138-5401