Learn More
Strem, An Ascensus Company CAS# 213697-53-1. 500mg. 2-(Dicyclohexylphosphino)-2'-(N,N-dimethylamino))-1,1'-biphenyl, 98% DavePhos. MFCD02183572
SDP
Catalog No. 5090110096
Encompass
CAS# 213697-53-1. 500mg. 2-(Dicyclohexylphosphino)-2'-(N,N-dimethylamino))-1,1'-biphenyl, 98% DavePhos. MFCD02183572. Molecular Weight: 393.55. Molecular Formula: C26H36NP. Color/form: white xtl. Strem# 15-1145. http://www.strem.com/catalog/v/15-1145/
Catalog No. 50-901-10096 Supplier Strem, An Ascensus Company Supplier No. 151145500MG
CAS# 213697-53-1. 500mg. 2-(Dicyclohexylphosphino)-2'-(N,N-dimethylamino))-1,1'-biphenyl, 98% DavePhos. MFCD02183572. Molecular Weight: 393.55. Molecular Formula: C26H36NP. Color/form: white xtl. Strem# 15-1145. http://www.strem.com/catalog/v/15-1145/