Learn More
Strem, An Ascensus Company CAS# 787618-22-8. 1g. 2-Dicyclohexylphosphino-2',6'-di-i-propoxy-1,1'-biphenyl, min. 98% RuPhos. MFCD06798294
SDP
Supplier:  Strem, An Ascensus Company 1511461G
Encompass
CAS# 787618-22-8. 1g. 2-Dicyclohexylphosphino-2',6'-di-i-propoxy-1,1'-biphenyl, min. 98% RuPhos. MFCD06798294. Molecular Weight: 466.64. Molecular Formula: C30H43O2P. Color/form: white pwdr. Strem# 15-1146. http://www.strem.com/catalog/v/15-1146/
Catalog No. NC0569887