Learn More
Strem, An Ascensus Company CAS# 870245-84-4. 2g. 2'-Dicyclohexylphosphino-2,6-di-i-propyl-4-sulfonato-1,1'-biphenyl hydrate sodium salt (XPhos-SO3Na). MFCD09265103
SDP
Catalog No. 5090110085
Encompass
CAS# 870245-84-4. 2g. 2'-Dicyclohexylphosphino-2,6-di-i-propyl-4-sulfonato-1,1'-biphenyl hydrate sodium salt (XPhos-SO3Na). MFCD09265103. Molecular Weight: 536.68. Molecular Formula: C30H42NaO3PS·;XH2O. Color/form: beige solid Strem# 15-1135. http://www.strem.com/catalog/v/15-1135/
Catalog No. 50-901-10085 Supplier Strem, An Ascensus Company Supplier No. 1511352G
CAS# 870245-84-4. 2g. 2'-Dicyclohexylphosphino-2,6-di-i-propyl-4-sulfonato-1,1'-biphenyl hydrate sodium salt (XPhos-SO3Na). MFCD09265103. Molecular Weight: 536.68. Molecular Formula: C30H42NaO3PS·;XH2O. Color/form: beige solid Strem# 15-1135. http://www.strem.com/catalog/v/15-1135/