Learn More
Medchemexpress LLC 2-(Dicyclohexylphosphino)-2',4',6'-triisopropylbiphenyl | 564483-18-7 | MFCD04117682 | 98.0% | 476.72 g·mol⁻¹ | C33H49P | 10g
SDP
Catalog No. 5000269393
Encompass_Preferred
XPhos is a dual aryl monophosphine ligand XPhos is an efficient and stable palladium catalytic ligand XPhos can be used for amination of aryl sulfonate esters[1][2]
Catalog No. 50-002-69393 Supplier Medchemexpress LLC Supplier No. HYY000610G
XPhos is a dual aryl monophosphine ligand XPhos is an efficient and stable palladium catalytic ligand XPhos can be used for amination of aryl sulfonate esters[1][2]