Learn More
Medchemexpress LLC 2-(dicyclohexylphosphino)-2',4',6'-triisopropylbiphenyl | 564483-18-7 | MFCD04117682 | 99.1% | 476.72 | C33H49P | 25g
SDP
Catalog No. 5000268264
Encompass_Preferred
XPhos is a dual aryl monophosphine ligand XPhos is an efficient and stable palladium catalytic ligand XPhos can be used for amination of aryl sulfonate esters[1][2]
Catalog No. 50-002-68264 Supplier Medchemexpress LLC Supplier No. HYY000625G
XPhos is a dual aryl monophosphine ligand XPhos is an efficient and stable palladium catalytic ligand XPhos can be used for amination of aryl sulfonate esters[1][2]