Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Diethylamino)ethanol Hydrochloride, TCI America™
SDP

Catalog No. D046725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D046725G 25 g
D0467500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D046725G Supplier TCI America Supplier No. D046725G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 14426-20-1
Molecular Formula C6H16ClNO
Molecular Weight (g/mol) 153.65
MDL Number MFCD00036374
InChI Key DBQUKJMNGUJRFI-UHFFFAOYSA-N
Synonym 2-diethylamino ethanol hydrochloride, dehydrasal, perdilaton, 2-diethylaminoethanol hydrochloride, ethanol, 2-diethylamino-, hydrochloride, deae hydrochloride, unii-4w7n0aa8f1, diethylaminoethanol hydrochloride, n,n-diethylethanolamine hydrochloride
PubChem CID 84429
IUPAC Name 2-(diethylamino)ethanol;hydrochloride
SMILES CCN(CC)CCO.Cl

Specifications

Melting Point 133°C
Color White
Quantity 25 g
Formula Weight 153.65
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Diethylamino)ethanol Hydrochloride
EINECSNumber (2)-0297&(2)-0353
RTECSNumber KK5553400
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.