Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Difluoromethoxy)benzaldehyde 98.0+%, TCI America™
SDP

Catalog No. D48371G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48371G 1 g
D48375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48371G Supplier TCI America Supplier No. D48371G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 71653-64-0
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.131
MDL Number MFCD00042251
InChI Key QPBNHDFPMRENBC-UHFFFAOYSA-N
Synonym 2-difluoromethoxy benzaldehyde, 2-difluoromethoxy-benzaldehyde, o-difluoromethoxybenzaldehyde, 2-difloromethoxybenzaldehyde, pubchem2906, acmc-209ojk, difluoromethoxybenzaldehyde, ksc356m9l, zerenex zx009675, benzaldehyde, difluoromethoxy
PubChem CID 2736984
IUPAC Name 2-(difluoromethoxy)benzaldehyde
SMILES C1=CC=C(C(=C1)C=O)OC(F)F

Specifications

Boiling Point 83°C
Quantity 1 g
Formula Weight 172.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Difluoromethoxy)benzaldehyde
RTECSNumber CU5615600
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.