Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(Dimethylamino)ethyl methacrylate

Catalog No. 2349071L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-490-71L 1 L
23-490-7100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-490-71L Supplier Sigma Aldrich Supplier No. 2349071L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-(Dimethylamino)ethyl methacrylate may be used in:the synthesis of poly ((2-(Dimethylamino)ethyl methacrylate)block copolymers of 2-(Dimethylamino)ethyl methacrylate and n-butyl methacrylateamphiphilic AB block copolymers of 2-(Dimethylamino)ethyl methacrylate

Specifications

CAS 2867-47-2
Density 0.933 g/mL (at 25°C)
Boiling Point 182°C to 192°C
Molecular Formula C8H15NO2
Refractive Index n20/D 1.439 (literature)
Linear Formula CH2=C(CH3)COOCH2CH2N(CH3)2
Quantity 1 L
MDL Number MFCD00008589
Synonym Methacrylic acid 2-(dimethylamino)ethyl ester
Molecular Weight (g/mol) 157.21
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.