Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Dimethylamino)ethyl methacrylate, 99%, stabilized

Catalog No. p-3736274
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC215842500 250 g Glass bottle
AC215840010 1 kg Glass bottle
AC215840051 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC215842500 Supplier Thermo Scientific Chemicals Supplier No. 215842500
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 2867-47-2 | C8H15NO2 | 157.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2867-47-2
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.21
MDL Number MFCD00008589
InChI Key JKNCOURZONDCGV-UHFFFAOYSA-N
Synonym 2-dimethylamino ethyl methacrylate, dimethylaminoethyl methacrylate, ageflex fm-1, dmaema, usaf rh-3, 2-propenoic acid, 2-methyl-, 2-dimethylamino ethyl ester, 2-n,n-dimethylamino ethyl methacrylate, n,n-dimethylaminoethyl methacrylate, 2-dimethylaminoethyl methacrylate, n,n-dimethylethanolamine methacrylate
PubChem CID 17869
IUPAC Name 2-(dimethylamino)ethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCN(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -50.0°C
Color Colorless to Yellow
Density 0.9330g/mL
Boiling Point 182.0°C to 192.0°C
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=C(CH3)CO2CH2CH2N(CH3)2
Refractive Index 1.4390 to 1.4410
Quantity 250 g
Beilstein 04, III, 649
Specific Gravity 0.933
Solubility Information Solubility in water: soluble.
Stabilizer 750 to 2000ppm MEHQ
Viscosity 1.32 mPa.s (20°C)
Formula Weight 157.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-(Dimethylamino)ethyl methacrylate, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 220-688-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.