Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Dimethylaminobenzoic acid, 98%

Catalog No. AAA1296706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1296706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1296706 Supplier Thermo Scientific Chemicals Supplier No. A1296706

CAS: 610-16-2 | C9H11NO2 | 165.192 g/mol

2-Dimethylaminobenzoic acid is used as a chemical and organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Dimethylaminobenzoic acid is used as a chemical and organic intermediate.

Solubility
Insoluble in water.

Notes
Store in tightly sealed containers in a cool place. Protect from moisture. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-16-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00016496
InChI Key DVVXXHVHGGWWPE-UHFFFAOYSA-N
Synonym 2-dimethylamino benzoic acid, n,n-dimethylanthranilic acid, benzoic acid, 2-dimethylamino, anthranilic acid, n,n-dimethyl, anthranilic acid,n-dimethyl, n,n-dimethylaminobenzoic acid, benzoic acid, dimethylamino, o-dimethylaminobenzoic acid, anthranilic acid, n,n-dimethyl-8ci, 2-dimethylamino-benzoic acid; hydrochloride
PubChem CID 69118
IUPAC Name 2-(dimethylamino)benzoic acid
SMILES CN(C)C1=CC=CC=C1C(=O)O

Specifications

Melting Point 70°C to 74°C
Odor Odorless
Quantity 5 g
Beilstein 2089466
Solubility Information Insoluble in water.
Formula Weight 165.19
Percent Purity 98%
Chemical Name or Material 2-Dimethylaminobenzoic acid
EINECSNumber 210-209-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.