Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Dimethylaminoethyl acetate, 98%, Thermo Scientific Chemicals

Catalog No. AC408061000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC408061000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC408061000 Supplier Thermo Scientific Chemicals Supplier No. 408061000

missing translation for 'validMainframeMsg'

CAS: 1421-89-2 | Molecular Formula: C6H13NO2 | Molecular Weight: 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1421-89-2
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00041912
InChI Key GOLSFPMYASLXJC-UHFFFAOYSA-N
Synonym 2-dimethylamino ethyl acetate, dimethylaminoethyl acetate, dimethylaminoethanol acetate, 2-dimethylaminoethanol acetate, acetic acid, 2-dimethylamino ethyl ester, acetic acid 2-dimethylaminoethyl ester, 2-dimethylamino ethanol acetic ester, acmc-1c2gl, 2-dimethylaminoethyl=acetate, 2-dimethylamino ethylacetate
PubChem CID 15013
IUPAC Name 2-(dimethylamino)ethyl acetate
SMILES CN(C)CCOC(C)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 0.92g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Refractive Index 1.4155 to 1.4175 (20°C, 589nm)
Linear Formula CH3COOCH2CH2N(CH3)2
Quantity 100 g
Formula Weight 131.17
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 2-Dimethylaminoethyl acetate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 215-827-4
RTECSNumber AH2100000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.