Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Dimethylvinylsilyl)pyridine 97.0+%, TCI America™
SDP

Catalog No. D29351G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D29351G Supplier TCI America Supplier No. D29351G

Chemical Identifiers

CAS 321155-39-9
Molecular Formula C9H13NSi
MDL Number MFCD06797092
Synonym 2-(Ethenyldimethylsilyl)pyridine, 2-(Vinyldimethylsilyl)pyridine

Specifications

Color Yellow
Quantity 1 g
Formula Weight 163.30
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Dimethylvinylsilyl)pyridine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.