Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-(+)-2-(Diphenylmethyl)Pyrrolidine

Catalog No. 5525421G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-254-21G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 55-254-21G Supplier Sigma Aldrich Supplier No. 5525421G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used as excellent chiral solvating agents to determine the enantiomeric composition of chiral carboxylic acids directly by NMR analysis.

Specifications

CAS 113826-06-5
Melting Point 46°C to 49°C
Molecular Formula C10H12O4S
Refractive Index n20/D 1.5870
Linear Formula C10H12O4S
MDL Number MFCD00010834
Quantity 25 g
Molecular Weight (g/mol) 228.26 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.