Learn More
eMolecules​ 65423-44-1 | 2-(Diphenylphosphino)aniline | Ambeed | MFCD12923152 | 277.307 | C18H16NP | 98 | Nc1ccccc1P(c1ccccc1)c1ccccc1 | 1g | 572829027
Catalog No. NC2293916 Shop All eMolecules Inc. Products
Encompass_Preferred
2-(Diphenylphosphino)aniline | Ambeed | 65423-44-1 | MFCD12923152 | 277.307 | C18H16NP | 98.000 | Nc1ccccc1P(c1ccccc1)c1ccccc1 | 1g | 572829027
This item is not returnable. View return policy
Catalog No. NC2293916 Supplier eMolecules​ Supplier No. 572829027
2-(Diphenylphosphino)aniline | Ambeed | 65423-44-1 | MFCD12923152 | 277.307 | C18H16NP | 98.000 | Nc1ccccc1P(c1ccccc1)c1ccccc1 | 1g | 572829027