Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Diphenylphosphino)benzoic acid, 97%

Catalog No. AAB2238903
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2238903 1 g
AAB2238906 5 g
AAB2238914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2238903 Supplier Thermo Scientific Chemicals Supplier No. B2238903
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 17261-28-8 | C19H15O2P | 306.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17261-28-8
Molecular Formula C19H15O2P
Molecular Weight (g/mol) 306.30
MDL Number MFCD00674024
InChI Key UYRPRYSDOVYCOU-UHFFFAOYSA-N
Synonym 2-diphenylphosphino benzoic acid, 2-diphenylphosphinobenzoic acid, benzoic acid, 2-diphenylphosphino, o-diphenylphosphinobenozic acid, 2-carboxyphenyl diphenylphosphine, 2-diphenylphosphanyl benzoic acid, benzoic acid, diphenylphosphino, o-diphenylphosphino benzoic acid, dppbac, acmc-1bt7u
PubChem CID 87021
IUPAC Name 2-diphenylphosphanylbenzoic acid
SMILES OC(=O)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 174°C to 181°C
Quantity 1 g
Formula Weight 306.31
Percent Purity 97%
Chemical Name or Material 2-(Diphenylphosphino)benzoic acid
Hazard Category H302+H332-H315-H319-H335-H500
Hazard Statement GHS H Statement
H332-H315-H319-H335
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 241-293-7
RTECSNumber DG9287500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.