Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Diphenylphosphino)ethylamine, 95%

Catalog No. AAH2707009
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2707009 10 g
AAH2707003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2707009 Supplier Thermo Scientific Chemicals Supplier No. H2707009
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4848-43-5 | C14H16NP | 229.26 g/mol

2-(Diphenylphosphino)ethylamine, is used as a fine chemical intermediate. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Diphenylphosphino)ethylamine, is used as a fine chemical intermediate. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Air Sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4848-43-5
Molecular Formula C14H16NP
Molecular Weight (g/mol) 229.26
MDL Number MFCD00233831
InChI Key RXEPBCWNHKZECN-UHFFFAOYSA-N
Synonym 2-diphenylphosphino ethylamine, 2-diphenylphosphino ethanamine, 2-aminoethyl diphenylphosphine, 1-amino-2-diphenylphosphino ethane, 2-aminoethyl diphenylphosphane, 2-diphenylphosphanyl ethan-1-amine, 2-diphenylphosphino ethanamine hydrochloride, acmc-209v7l, nh2 ch2 2pph2, 2-diphenylphosphino-ethylamine
PubChem CID 273264
IUPAC Name 2-diphenylphosphanylethanamine
SMILES NCCP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 10 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 229.27
Percent Purity 95%
Chemical Name or Material 2-(Diphenylphosphino)ethylamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.