Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(Diphenylphosphino)pyridine | 37943-90-1 | ≥97.0% | 263.28 | 10 G
SDP

Catalog No. 5000454647
Encompass_Preferred

Diphenyl-2-pyridylphosphine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is a ligand for metal-catalyzed carbonylations, hydration, dehydrogenative coupling, carbostannylation, distannylation, and silylation. It is also employed as a reagent for Mitsunobu reaction.

  • Appearance: solid
  • Color: white to light yellow
  • Shipping: room temperature in continental US; may vary elsewhere.
  • Storage: 4°C, stored under nitrogen. In solvent: -80°C, 6 months; -20°C, 1 month (stored under nitrogen).

Catalog No. 50-004-54647 Supplier Medchemexpress LLC Supplier No. HYW01334310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diphenyl-2-pyridylphosphine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is a ligand for metal-catalyzed carbonylations, hydration, dehydrogenative coupling, carbostannylation, distannylation, and silylation. It is also employed as a reagent for Mitsunobu reaction.

  • Appearance: solid
  • Color: white to light yellow
  • Shipping: room temperature in continental US; may vary elsewhere.
  • Storage: 4°C, stored under nitrogen. In solvent: -80°C, 6 months; -20°C, 1 month (stored under nitrogen).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.