Learn More
Julabo USA Inc Pair, Twin adapter G 1 1/4" female to 2x 1" hose barbs
SDP
Catalog No. 507534229
Encompass_Preferred
Bath Accessories, Julabo, Twin distributing adapter G1 1/4″ female fitting with 2 barbed fittings for tubing 1″ ID. Suitable for use with 6 bar pump FL chillers: FL(W)7006, FL(W)11006, FL(W)20006. Stainless steel.
Catalog No. 50-753-4229 Supplier Julabo USA Inc Supplier No. 8970477
Bath Accessories, Julabo, Twin distributing adapter G1 1/4″ female fitting with 2 barbed fittings for tubing 1″ ID. Suitable for use with 6 bar pump FL chillers: FL(W)7006, FL(W)11006, FL(W)20006. Stainless steel.