Learn More
Julabo USA Inc Pair, Twin adapter G 3/4" female to 2x 3/4" hose barbs
SDP
Catalog No. 507534228
Encompass_Preferred
Bath Accessories, Julabo, Twin distributing adapter G 3/4″ female fitting with 2x barbed fittings for tubing 3/4″ ID. Suitable for use with 3 bar pump FL chillers: FL(W)4003, FL(W)2503. Stainless steel.
Catalog No. 50-753-4228 Supplier Julabo USA Inc Supplier No. 8970476
Bath Accessories, Julabo, Twin distributing adapter G 3/4″ female fitting with 2x barbed fittings for tubing 3/4″ ID. Suitable for use with 3 bar pump FL chillers: FL(W)4003, FL(W)2503. Stainless steel.