Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Dodecanone, 95%

Catalog No. AAA1886218
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1886218 50 g
AAA1886209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1886218 Supplier Thermo Scientific Chemicals Supplier No. A1886218
Add to Cart

CAS: 6175-49-1 | C12H24O | 184.323 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6175-49-1
Molecular Formula C12H24O
Molecular Weight (g/mol) 184.323
MDL Number MFCD00015064
InChI Key LSKONYYRONEBKA-UHFFFAOYSA-N
Synonym 2-dodecanone, methyl decyl ketone, decyl methyl ketone, unii-p5cn8ysv3p, dodecanone-2, p5cn8ysv3p, n-decyl methyl ketone, dodecanone, acmc-1b2ve, 2-dodecanone gc
PubChem CID 22556
IUPAC Name dodecan-2-one
SMILES CCCCCCCCCCC(=O)C

Specifications

Melting Point 17°C to 20°C
Density 0.82
Boiling Point 247°C
Flash Point 110°C (230°F)
Refractive Index 1.433
Quantity 50 g
Beilstein 1703135
Formula Weight 184.32
Percent Purity ≥98%
Chemical Name or Material 2-Dodecanone
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 228-222-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.