Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2-Dodecen-1-yl)succinic anhydride | 19780-11-1 | ≥97.0% | 266.38 | 1 ML
SDP

Catalog No. 5000451511
Encompass_Preferred

(2-Dodecen-1-yl)succinic anhydride is an organic compound used in certain polymerization reactions for manufacturing plastics and resins. It is also utilized in industrial fields such as coatings, inks, and textile manufacturing, and serves as a raw material for pharmaceutical intermediates and surfactants. It is a biochemical reagent for life science research.

  • Appearance: Solid
  • Color: White to off-white
  • Formula: C16H26O3
  • Shipping: Room temperature in continental US
  • Storage: 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light).
  • Solubility (In Vitro): DMSO: 250 mg/mL (938.51 mM)

Catalog No. 50-004-51511 Supplier Medchemexpress LLC Supplier No. HYW00940010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(2-Dodecen-1-yl)succinic anhydride is an organic compound used in certain polymerization reactions for manufacturing plastics and resins. It is also utilized in industrial fields such as coatings, inks, and textile manufacturing, and serves as a raw material for pharmaceutical intermediates and surfactants. It is a biochemical reagent for life science research.

  • Appearance: Solid
  • Color: White to off-white
  • Formula: C16H26O3
  • Shipping: Room temperature in continental US
  • Storage: 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light).
  • Solubility (In Vitro): DMSO: 250 mg/mL (938.51 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.