Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2-Dodecen-1-yl)succinic anhydride | 19780-11-1 | ≥97.0% | 266.38 | 500 MG
SDP

Catalog No. 5000366869
Encompass_Preferred

(2-Dodecen-1-yl)succinic anhydride is an organic compound utilized in polymerization reactions and various industrial applications. It serves in the manufacture of plastics, resins, coatings, inks, and in textile manufacturing. This compound also functions as a raw material for pharmaceutical intermediates and surfactants.

  • Used in polymerization reactions
  • Raw material for plastics, resins, coatings, and inks
  • Used in textile manufacturing
  • Raw material for pharmaceutical intermediates and surfactants
  • Purity ≥97.0%
  • Molecular weight 266.38
  • Solid, white to off-white appearance
  • Soluble in DMSO (250 mg/mL)
  • Storage at 4 °C, protected from light
  • Melting point 41-43 °C
  • Boiling point 180-182 °C at 5 mmHg
  • Relative density 1.00 g/cm³

Catalog No. 50-003-66869 Supplier Medchemexpress LLC Supplier No. HYW009400500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(2-Dodecen-1-yl)succinic anhydride is an organic compound utilized in polymerization reactions and various industrial applications. It serves in the manufacture of plastics, resins, coatings, inks, and in textile manufacturing. This compound also functions as a raw material for pharmaceutical intermediates and surfactants.

  • Used in polymerization reactions
  • Raw material for plastics, resins, coatings, and inks
  • Used in textile manufacturing
  • Raw material for pharmaceutical intermediates and surfactants
  • Purity ≥97.0%
  • Molecular weight 266.38
  • Solid, white to off-white appearance
  • Soluble in DMSO (250 mg/mL)
  • Storage at 4 °C, protected from light
  • Melting point 41-43 °C
  • Boiling point 180-182 °C at 5 mmHg
  • Relative density 1.00 g/cm³

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.