Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(Dodecylthiocarbonothioylthio)-2-methylpropionic acid 3-azido-1-propanol ester

Catalog No. 7416985G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-169-85G 5 g
74-169-81G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 74-169-85G Supplier Sigma Aldrich Supplier No. 7416985G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Need help choosing the correct RAFT Agent? Please consult the RAFT Agent to Monomer compatibility table.Functionalized RAFT agent for controlled radical polymerization; especially suited for the polymerization of styrene, acrylate and acrylamide monomers. Azide group can be used to conjugate to a variety of alkyne-functionalized biomolecules. Chain Transfer Agent (CTA).

Specifications

CAS 927816-03-3
Density 1.058 g/mL (at 25°C)
Molecular Formula C20H37N3O2S3
Linear Formula C20H37N3O2S3
Refractive Index n20/D 1.528
MDL Number MFCD22200504
Quantity 5 g
Molecular Weight (g/mol) 447.72
Percent Purity 98% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.