Learn More
Fire King Security Group 2 drw, 1 hr fire & impact rated, 44 in wide lateral Black Finish
SDP
Catalog No. NC2952289
Encompass_Preferred
Two-drawer lateral 44 in wide UL Class 350 one-hour fire / impact rated filing cabinet with one high-security UL listed keylock top drawer to control all drawers. ETL Verified Mark 1/2 hour fire protection hard drives, CDs, DVDs, flash drives.
Catalog No. NC2952289 Supplier Fire King Security Group Supplier No. 24422CBL
Add to Cart
Add to Cart
Request a Quote
Two-drawer lateral 44 in wide UL Class 350 one-hour fire / impact rated filing cabinet with one high-security UL listed keylock top drawer to control all drawers. ETL Verified Mark 1/2 hour fire protection hard drives, CDs, DVDs, flash drives.