Learn More
Analytical Sales & Services 2 DRAM, 24-Position Parallel Synthesis Reaction Block, Gen II
Catalog No. 5000806034
Encompass
Gen II, 24 Well, 5-BOLT, Parallel Synthesis Reaction Block\n• For 17mm OD (2 Dram) Vials\n• 18mm Well Spacing\n• Includes Vial Rack, Top Cover, PFA Film, Rubber Mats and Screws (vials not included)
Catalog No. 50-008-06034 Supplier Analytical Sales & Services Supplier No. 106240
Gen II, 24 Well, 5-BOLT, Parallel Synthesis Reaction Block\n• For 17mm OD (2 Dram) Vials\n• 18mm Well Spacing\n• Includes Vial Rack, Top Cover, PFA Film, Rubber Mats and Screws (vials not included)