Learn More
Burch Bottle & Packaging Inc 2 DRAM FLINT VIAL 15/425
Catalog No. NC1829606
Encompass
Stock number 1GV250002GV is a 2 Dram Glass Vial with a 15-425 continuous thread finish. It is made out of type 1 Borosilicate, giving it a high-quality feel and a crystal clear finish. The 2 Dram Glass Vial is tall and narrow and is the ideal packaging option for labware and pharmaceutical products due to its high chemical resistance. It's also perfect for essential oils or solvents.
Catalog No. NC1829606 Supplier Burch Bottle & Packaging Inc Supplier No. 1GV250002GV
Stock number 1GV250002GV is a 2 Dram Glass Vial with a 15-425 continuous thread finish. It is made out of type 1 Borosilicate, giving it a high-quality feel and a crystal clear finish. The 2 Dram Glass Vial is tall and narrow and is the ideal packaging option for labware and pharmaceutical products due to its high chemical resistance. It's also perfect for essential oils or solvents.