Learn More
Ace Glass, Inc. 2 dram poly-seal vial, cs/1728, sp/144
SDP
Catalog No. 50976409
Encompass
2 dram sample vials fabricated from borosilicate glass. Include poly-seal cone-lined closures, which are molded in a premium grade, low density polyethylene. Inverted cone liner provides excellent seal and torque retention.
Catalog No. 50-976-409 Supplier Ace Glass, Inc. Supplier No. 877920
2 dram sample vials fabricated from borosilicate glass. Include poly-seal cone-lined closures, which are molded in a premium grade, low density polyethylene. Inverted cone liner provides excellent seal and torque retention.