Learn More
USA Scientific Inc 2-drawer rack for 15 mL tubes, 120 tube capacity
Catalog No. 507541170
Encompass
2-drawer upright freezer rack for 15 mL tubes, 120 tube capacity, 16 1/2 in L x 5 1/2 in W x 9 15/16 in H, stainless steel
Catalog No. 50-754-1170 Supplier USA Scientific Inc Supplier No. 24833815
2-drawer upright freezer rack for 15 mL tubes, 120 tube capacity, 16 1/2 in L x 5 1/2 in W x 9 15/16 in H, stainless steel