Learn More
Medchemexpress LLC 2-[[(E)-3-(4-hydroxyphenyl)prop-2-enoyl]amino]benzoic acid | 115610-36-1 | MFCD13554294 | 99.9% | 283.28 g/mol | C16H13NO4 | 25 MG
SDP
Catalog No. 5000229118
Encompass_Preferred
Avenanthramide D is a naturally occurring phenolic acid isolated from Avena sativa L., supplied for research applications investigating inflammation and related biochemical pathways.
  • High purity suitable for analytical and biological research (>99.8%).
  • Molecular formula C16H13NO4; molecular weight 283.28 g/mol.
  • Offered in small laboratory quantities such as 25 MG for convenience in assays.
  • Store protected from light at 4°C; in solvent, store at -80°C for long-term stability.
  • Applicable to studies of anti-inflammatory activity and phenolic acid biochemistry.
Catalog No. 50-002-29118 Supplier Medchemexpress LLC Supplier No. HY11869325MG
Avenanthramide D is a naturally occurring phenolic acid isolated from Avena sativa L., supplied for research applications investigating inflammation and related biochemical pathways.
  • High purity suitable for analytical and biological research (>99.8%).
  • Molecular formula C16H13NO4; molecular weight 283.28 g/mol.
  • Offered in small laboratory quantities such as 25 MG for convenience in assays.
  • Store protected from light at 4°C; in solvent, store at -80°C for long-term stability.
  • Applicable to studies of anti-inflammatory activity and phenolic acid biochemistry.