Learn More
Cayman Chemical 2 E 4 ZDecadIenoIc AcId et 10g
SDP
Catalog No. 502988252
Encompass_Preferred
A volatile compound with kairomonal attractant activity; attracts male and female codling moths (C. pomonella) to traps placed in orchards at 40 mg/trap; attracts C. splendana males and females, as well as P. fasciana males
Catalog No. 50-298-8252 Supplier Cayman Chemical Supplier No. 3083510 G
A volatile compound with kairomonal attractant activity; attracts male and female codling moths (C. pomonella) to traps placed in orchards at 40 mg/trap; attracts C. splendana males and females, as well as P. fasciana males