Learn More
Cayman Chemical 2(E),4(Z)-Decadienoic Acid ethyl ester, Purity: ≥95%, CAS: 3025-30-7, 50 g
SDP
Catalog No. 502989825
Encompass_Preferred
A volatile compound with kairomonal attractant activity; attracts male and female codling moths (C. pomonella) to traps placed in orchards at 40 mg/trap; attracts C. splendana males and females, as well as P. fasciana males
Catalog No. 50-298-9825 Supplier Cayman Chemical Supplier No. 3083550 G
A volatile compound with kairomonal attractant activity; attracts male and female codling moths (C. pomonella) to traps placed in orchards at 40 mg/trap; attracts C. splendana males and females, as well as P. fasciana males