Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-[(E)-[(5-bromothiophen-2-yl)methylidene]amino]benzamide | 304456-62-0 | MFCD00977636 | 98.0% | 309.18 g/mol | C12H9BrN2OS | 250MG
SDP

Catalog No. 5000235587
Encompass_Preferred

BTYNB is a research-grade small-molecule inhibitor of the RNA-binding protein IMP1 (IGF2BP1) that disrupts IMP1 binding to c-Myc mRNA (reported IC50 = 5 μM). It is supplied as a characterized research reagent for laboratory studies and is not for human or clinical use.

  • Potent inhibitor of IMP1 binding to c-Myc mRNA (IC50 = 5 μM).
  • High reported purity, 98.0%.
  • Molecular weight 309.18 g/mol.
  • Chemical formula C12H9BrN2OS.
  • Provided as a 250 mg research reagent.
  • Intended for laboratory research use only.

Catalog No. 50-002-35587 Supplier Medchemexpress LLC Supplier No. HY124447250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BTYNB is a research-grade small-molecule inhibitor of the RNA-binding protein IMP1 (IGF2BP1) that disrupts IMP1 binding to c-Myc mRNA (reported IC50 = 5 μM). It is supplied as a characterized research reagent for laboratory studies and is not for human or clinical use.

  • Potent inhibitor of IMP1 binding to c-Myc mRNA (IC50 = 5 μM).
  • High reported purity, 98.0%.
  • Molecular weight 309.18 g/mol.
  • Chemical formula C12H9BrN2OS.
  • Provided as a 250 mg research reagent.
  • Intended for laboratory research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.