Learn More
Electron Microscopy Sciences 2-Etched 15 mm Circle Slide
SDP
Catalog No. 50949722
Encompass_Preferred
Useful for fluorescent antibody examination. The size of the slide is 3"x1", approximately 1 mm thick, with an etched frosted end. Pre-cleaned. Packed 72/box.
Catalog No. 50-949-722 Supplier Electron Microscopy Sciences Supplier No. 7187916
Useful for fluorescent antibody examination. The size of the slide is 3"x1", approximately 1 mm thick, with an etched frosted end. Pre-cleaned. Packed 72/box.