Learn More
AdipoGen 2-Ethoxy-2-2-naphthyl-MeCN
SDP
Supplier:  AdipoGen CDXE0031M250
Encompass_Preferred
Chemical. CAS 33224-80-5. Formula C14H13NO. MW 211.26. Synthetic. This is a new substrate for Cytochrom P450 suitable for fluorescence assays.
Catalog No. 50-256-0991