Learn More
Medchemexpress LLC 2-Ethoxybenzamide | 938-73-8 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB1428500MG
Encompass_Preferred
2-Ethoxybenzamide (Standard) is an analytical standard used as a reference in assays. It is a nonsteroidal anti-inflammatory agent with analgesic and antipyretic effects. It induces melanin synthesis via cAMP response element-binding protein (CREB) phosphorylation and can be used in research for hypopigmentation and inflammation-related diseases.
  • Used in qualitative research experiments.
  • Used in quantitative research experiments.
  • Used in methodological research experiments.
  • Suitable for HPLC, GC, and MS applications.
Catalog No. 50-004-13489