Learn More
Medchemexpress LLC 2-Ethoxybenzamide | 938-73-8 | >99.8% | 1 ML
SDP
Catalog No. 5000413005
Encompass_Preferred
2-Ethoxybenzamide (Ethenzamide) is a nonsteroidal anti-inflammatory agent with analgesic and antipyretic effects. It induces melanin synthesis through cAMP response element-binding protein (CREB) phosphorylation and can be used in research for hypopigmentation and inflammation-related diseases. In B16F1 murine melanoma cells, it increases ROS levels, enhances the expression of melanogenesis-related genes, and promotes melanin synthesis via the phosphorylation of ERK/CREB.
  • Analgesic and antipyretic effects
  • Induces melanin synthesis via CREB phosphorylation
  • Useful in research for hypopigmentation and inflammation-related diseases
  • Increases ROS levels in B16F1 murine melanoma cells
  • Enhances melanogenesis-related gene expression
Catalog No. 50-004-13005 Supplier Medchemexpress LLC Supplier No. HYB142810MM/1ML
2-Ethoxybenzamide (Ethenzamide) is a nonsteroidal anti-inflammatory agent with analgesic and antipyretic effects. It induces melanin synthesis through cAMP response element-binding protein (CREB) phosphorylation and can be used in research for hypopigmentation and inflammation-related diseases. In B16F1 murine melanoma cells, it increases ROS levels, enhances the expression of melanogenesis-related genes, and promotes melanin synthesis via the phosphorylation of ERK/CREB.
  • Analgesic and antipyretic effects
  • Induces melanin synthesis via CREB phosphorylation
  • Useful in research for hypopigmentation and inflammation-related diseases
  • Increases ROS levels in B16F1 murine melanoma cells
  • Enhances melanogenesis-related gene expression