Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxybenzenesulfonyl chloride, 96%

Catalog No. AAH3432506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3432506 5 g
AAH3432503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3432506 Supplier Thermo Scientific Chemicals Supplier No. H3432506
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 68800-33-9 | C8H9ClO3S | 220.667 g/mol

2-Ethoxybenzenesulfonyl chloride is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethoxybenzenesulfonyl chloride is used as pharmaceutical intermediates.

Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, water/ moisture, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68800-33-9
Molecular Formula C8H9ClO3S
Molecular Weight (g/mol) 220.667
MDL Number MFCD08706420
InChI Key RWURYJNJYODVAL-UHFFFAOYSA-N
Synonym 2-ethoxy-benzenesulfonyl chloride, 2-ethoxybenzene-1-sulfonyl chloride, 2-ethoxyphenylsulfonyl chloride, benzenesulfonylchloride, 2-ethoxy
PubChem CID 18944993
IUPAC Name 2-ethoxybenzenesulfonyl chloride
SMILES CCOC1=CC=CC=C1S(=O)(=O)Cl

Specifications

Melting Point 60°C to 64°C
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 220.67
Percent Purity 96%
Chemical Name or Material 2-Ethoxybenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.