Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(ethoxycarbonyl)-3-methylbutanoic acid | 15112-52-4 | 174.19 g/mol | C8H14O4 | 10 MG
SDP

Catalog No. 5000297863
Encompass_Preferred

2-(Ethoxycarbonyl)-3-methylbutanoic acid is a small-molecule organic intermediate used in the synthesis of active pharmaceutical compounds and research chemicals. It is intended for laboratory research use only and is offered in small pack sizes suitable for synthetic chemistry and method development.

  • Used as a drug intermediate in organic synthesis.
  • Suitable for method development and small-scale synthesis.
  • Molecular formula C8H14O4; molecular weight 174.19 g/mol.
  • Purity documentation is available from the supplier.

Catalog No. 50-002-97863 Supplier Medchemexpress LLC Supplier No. HY4696810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(Ethoxycarbonyl)-3-methylbutanoic acid is a small-molecule organic intermediate used in the synthesis of active pharmaceutical compounds and research chemicals. It is intended for laboratory research use only and is offered in small pack sizes suitable for synthetic chemistry and method development.

  • Used as a drug intermediate in organic synthesis.
  • Suitable for method development and small-scale synthesis.
  • Molecular formula C8H14O4; molecular weight 174.19 g/mol.
  • Purity documentation is available from the supplier.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.