Learn More
Medchemexpress LLC 2-(Ethoxycarbonyl)-3-methylbutanoic acid | 15112-52-4 | C8H14O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY46968
Encompass_Preferred
2-(Ethoxycarbonyl)-3-methylbutanoic acid, also known as 2-Isopropyl-malonic acid monoethyl ester, is a chemical compound primarily used as a drug intermediate for the synthesis of various active compounds. It is intended for research use only.
  • Drug intermediate for synthesis of various active compounds
  • Research use only
  • Oil appearance
  • Store at 4°C, protected from light and under nitrogen
  • In solvent: store at -80°C for 6 months; -20°C for 1 month, protected from light and under nitrogen
Catalog No. 50-003-58992