Learn More
Sigma Aldrich Fine Chemicals Biosciences 2-Ethoxyethanol ReagentPlu1L
Catalog No. 502898170
Encompass
2-Ethoxyethanol is a glycol ether. It has been reported as a precursor for ethoxyacetic acid (EAA). Various physical properties (density refractive index viscosity and speed of sound) of its binary mixtures with n-alkanes 2 2 4-trimethylpentane and cyclohexane have been evaluated. High-pressure vapor-liquid equilibrium (VLE) data of its binary mixtures with CO2 have been reported. It can be used as a solvent in the spectrophotometric determination of iron(III) by using the ligand N N-bis(2-hydroxy-5-bromo-benzyl)1 2 diaminopropane (HBDAP).
Catalog No. 50-289-8170 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1280821L
2-Ethoxyethanol is a glycol ether. It has been reported as a precursor for ethoxyacetic acid (EAA). Various physical properties (density refractive index viscosity and speed of sound) of its binary mixtures with n-alkanes 2 2 4-trimethylpentane and cyclohexane have been evaluated. High-pressure vapor-liquid equilibrium (VLE) data of its binary mixtures with CO2 have been reported. It can be used as a solvent in the spectrophotometric determination of iron(III) by using the ligand N N-bis(2-hydroxy-5-bromo-benzyl)1 2 diaminopropane (HBDAP).