Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxynaphthoic acid, 98%, Thermo Scientific™

Catalog No. AC243000500
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC243000500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC243000500 Supplier Thermo Scientific Chemicals Supplier No. 243000500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2224-00-2
Molecular Formula C13H11O3
Molecular Weight (g/mol) 215.23
MDL Number MFCD00004008
InChI Key MYFBSSDLYGWAHH-UHFFFAOYSA-M
Synonym 2-ethoxy-1-naphthoic acid, 2-ethoxynaphthoic acid, 1-naphthalenecarboxylic acid, 2-ethoxy, 2-ethoxy-1-naphthoicacid, 2-ethoxynaphthalenecarboxylic acid, acmc-209ftv, rarechem al be 0566, timtec-bb sbb005771, 2-ethoxynaphtoic acid
PubChem CID 75210
SMILES CCOC1=CC=C2C=CC=CC2=C1C([O-])=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 141°C to 145°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 50 g
IUPAC Name 2-ethoxynaphthalene-1-carboxylate
Formula Weight 216.24
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 2-Ethoxynaphthoic acid, 98%
Show More Show Less
EINECSNumber 218-745-7
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.