Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Ethyl-2-hydroxybutanoic acid | 3639-21-2 | MFCD00004183 | 98.0% | C6H12O3 | 10 MM * 1 ML
SDP

Catalog No. 5000353625
Encompass_Preferred

2-Ethyl-2-hydroxybutanoic acid is a hydroxy fatty acid and a metabolite of DEHP.

  • Hydroxy fatty acid
  • Metabolite of DEHP
  • Molecular weight: 132.16
  • Formula: C6H12O3
  • CAS number: 3639-21-2
  • Appearance: Solid
  • Color: White to off-white
  • SMILES: CCC(O)(CC)C(O)=O
  • Structure classification: Ketones, aldehydes, acids
  • Initial source: Endogenous metabolite

Catalog No. 50-003-53625 Supplier Medchemexpress LLC Supplier No. HY14210510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Ethyl-2-hydroxybutanoic acid is a hydroxy fatty acid and a metabolite of DEHP.

  • Hydroxy fatty acid
  • Metabolite of DEHP
  • Molecular weight: 132.16
  • Formula: C6H12O3
  • CAS number: 3639-21-2
  • Appearance: Solid
  • Color: White to off-white
  • SMILES: CCC(O)(CC)C(O)=O
  • Structure classification: Ketones, aldehydes, acids
  • Initial source: Endogenous metabolite

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.