Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethyl-2-(Hydroxymethyl) 1,3-Propanediol, Spectrum™ Chemical
SDP

Catalog No. 18620717 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
12 kg
50 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-717 500 g
18-620-716 25 g
18-620-718 2.5 kg
18-620-719 12 kg
18-620-720 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-620-717 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E2083500GM
Add to Cart
Add to Cart

Synonym: Adipic Acid, Trimethylolpropane, 1,6-Hexanediol

Chemical Identifiers

CAS 77-99-6

Specifications

Boiling Point 160°C (320°F)
Quantity 500 g
Molecular Weight (g/mol) 134.18
Percent Purity 99%
Grade Ungraded
Physical Form Powder or Waxy Solid
Chemical Name or Material 2-Ethyl-2-(hydroxymethyl) 1,3-Propanediol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.