Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethyl-2-imidazoline 98.0+%, TCI America™
SDP

Catalog No. E02345G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E02345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E02345G Supplier TCI America Supplier No. E02345G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 930-52-9
Molecular Formula C5H10N2
Molecular Weight (g/mol) 98.15
MDL Number MFCD00242607
InChI Key QUPKCFBHJFNUEW-UHFFFAOYSA-N
PubChem CID 13590
IUPAC Name 2-ethyl-4,5-dihydro-1H-imidazole
SMILES CCC1=NCCN1

Specifications

Melting Point 44°C
Color White-Yellow
Boiling Point 120°C
Quantity 5 g
Formula Weight 98.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Ethyl-2-imidazoline
EINECSNumber (5)-0422
RTECSNumber NJ3050000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.