Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethyl-3,5(6)-dimethylpyrazine, 99%, mixture of isomers, Thermo Scientific Chemicals

Catalog Number AAH2738306
Click to view available options
Quantity:
5 g
25 g
13925-07-0
C8H12N2
136.198
MFCD00047392
JZBCTZLGKSYRSF-UHFFFAOYSA-N
3,5-dimethyl-2-ethylpyrazine, pyrazine, 2-ethyl-3,5-dimethyl, 2,6-dimethyl-3-ethylpyrazine, 3-ethyl-2,6-dimethylpyrazine, 2-ethyl-3,5-dimethyl pyrazine, pyrazine, 2,6-dimethyl-3-ethyl, unii-r182ey5l8c, fema no. 3149, 2-ethyl-3,5 6-dimethylpyrazine, pyrazine, 3,5-dimethyl-2-ethyl
26334
2-ethyl-3,5-dimethylpyrazine
CCC1=NC=C(N=C1C)C
13925-07-0
C8H12N2
136.198
MFCD00047392
JZBCTZLGKSYRSF-UHFFFAOYSA-N
3,5-dimethyl-2-ethylpyrazine, pyrazine, 2-ethyl-3,5-dimethyl, 2,6-dimethyl-3-ethylpyrazine, 3-ethyl-2,6-dimethylpyrazine, 2-ethyl-3,5-dimethyl pyrazine, pyrazine, 2,6-dimethyl-3-ethyl, unii-r182ey5l8c, fema no. 3149, 2-ethyl-3,5 6-dimethylpyrazine, pyrazine, 3,5-dimethyl-2-ethyl
26334
2-ethyl-3,5-dimethylpyrazine
CCC1=NC=C(N=C1C)C

It is applied as an odorant of coffee brews, nutty flavor products and roasted food products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as an odorant of coffee brews, nutty flavor products and roasted food products.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol, chloroform and dichloromethane.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Density 0.96
Boiling Point 182°C
Flash Point 66°C (151°F)
Refractive Index 1.5015
Quantity 5 g
Solubility Information Not miscible or difficult to mix in water. Soluble in alcohol,chloroform and dichloromethane.
Formula Weight 136.19
Percent Purity 99%
Chemical Name or Material 2-Ethyl-3,5(6)-dimethylpyrazine, mixture of isomers
Hazard Category H227-H302
Hazard Statement GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.
Precautionary Statement P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c
EINECSNumber 237-694-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit