Learn More
Medchemexpress LLC 2-Ethyl-3-hydroxy-4H-pyran-4-one (Standard) | 4940-11-8 | 100.0% | 140.14 | 250 MG
SDP
Catalog No. 5000366408
Encompass_Preferred
Ethyl maltol (Standard) serves as an analytical standard for research and analytical applications. This compound, also known as 2-Ethyl-3-hydroxy-4H-pyran-4-one, is an orally active and significant food additive and flavor enhancer. It has been observed to be less toxic to rats and dogs.
  • Intended for research and analytical applications
  • Functions as an orally active food additive and flavor enhancer
  • Less toxic to rats and dogs
  • Enhances copper-mediated cytotoxicity
  • Induces apoptosis in lung epithelial cells
  • Solid appearance
  • Color: white to off-white
Catalog No. 50-003-66408 Supplier Medchemexpress LLC Supplier No. 250MGHYW010320R
Ethyl maltol (Standard) serves as an analytical standard for research and analytical applications. This compound, also known as 2-Ethyl-3-hydroxy-4H-pyran-4-one, is an orally active and significant food additive and flavor enhancer. It has been observed to be less toxic to rats and dogs.
  • Intended for research and analytical applications
  • Functions as an orally active food additive and flavor enhancer
  • Less toxic to rats and dogs
  • Enhances copper-mediated cytotoxicity
  • Induces apoptosis in lung epithelial cells
  • Solid appearance
  • Color: white to off-white